ENA308584592 Enamine

[5-(2-chlorophenyl)-1,3,4-oxadiazol-2-yl]methanol

  • MDL number MFCD08443044
  • Cas Number : 54014-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69355

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69355
Availability details for
Quantity
  • Availability
  • Add to Cart