ENA308584596 Enamine

3-phenylpentanoic acid

  • MDL number MFCD01722151
  • Cas Number : 5669-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61-62
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69359

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69359
Availability details for
Quantity
  • Availability
  • Add to Cart