ENA308584598 Enamine

1-(3-cyanopyridin-2-yl)piperidine-4-carboxamide

  • MDL number MFCD03086333
  • Cas Number : 700815-33-4
  • Molecular Weight : 230.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69093

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69093
Availability details for
Quantity
  • Availability
  • Add to Cart