ENA308584600 Enamine

2-(2-methyl-2,3-dihydro-1H-indol-1-yl)benzoic acid

  • MDL number MFCD12736546
  • Cas Number : 72990-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69363

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69363
Availability details for
Quantity
  • Availability
  • Add to Cart