ENA308584604 Enamine

4-hydroxyoxane-4-carboxylic acid

  • MDL number MFCD14602031
  • Cas Number : 50289-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69368

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69368
Availability details for
Quantity
  • Availability
  • Add to Cart