ENA308584606 Enamine

3-[2-(trifluoromethyl)phenyl]-1,2-oxazol-5-amine

  • MDL number MFCD07383045
  • Cas Number : 119162-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69051

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69051
Availability details for
Quantity
  • Availability
  • Add to Cart