ENA308584608 Enamine

(3,4-difluorophenyl)(pyridin-3-yl)methanamine

  • MDL number MFCD09026475
  • Cas Number : 939757-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-69372

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69372
Availability details for
Quantity
  • Availability
  • Add to Cart