ENA308584609 Enamine

N-[4-(2-aminoethyl)phenyl]acetamide

  • MDL number MFCD06213247
  • Cas Number : 63630-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121...123
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69373

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-69373
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart