ENA308584610 Enamine

4-amino-N-cyclopropyl-N-methylbenzene-1-sulfonamide

  • MDL number MFCD11623797
  • Cas Number : 1095038-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69375

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69375
Availability details for
Quantity
  • Availability
  • Add to Cart