ENA308584611 Enamine

2-(2-oxo-1,3-oxazinan-3-yl)-3-phenylpropanoic acid

  • MDL number MFCD13430406
  • Cas Number : 1258639-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69376

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69376
Availability details for
Quantity
  • Availability
  • Add to Cart