ENA308584612 Enamine

N,N-dimethyl-4-{[(2,2,2-trifluoroethyl)amino]methyl}aniline

  • MDL number MFCD09938951
  • Cas Number : 1016811-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69377

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69377
Availability details for
Quantity
  • Availability
  • Add to Cart