ENA308584613 Enamine

1-[1-(2,4-difluorophenyl)ethyl]piperazine

  • MDL number MFCD05187878
  • Cas Number : 518972-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69378

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69378
Availability details for
Quantity
  • Availability
  • Add to Cart