ENA308584615 Enamine

4-{[(cyclopropylmethyl)amino]methyl}-N,N-dimethylaniline

  • MDL number MFCD11140253
  • Cas Number : 1019611-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69380

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69380
Availability details for
Quantity
  • Availability
  • Add to Cart