ENA308584617 Enamine

N-(3-methoxypropyl)-2-(piperazin-1-yl)propanamide

  • MDL number MFCD10011194
  • Cas Number : 1018252-09-9
  • Molecular Weight : 229.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69382

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69382
Availability details for
Quantity
  • Availability
  • Add to Cart