ENA308584619 Enamine

2-(1-aminoethyl)-4-methoxyphenol

  • MDL number MFCD09047387
  • Cas Number : 926271-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69384

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69384
Availability details for
Quantity
  • Availability
  • Add to Cart