ENA308584620 Enamine

2-(dimethylamino)-2-(4-fluorophenyl)propanoic acid

  • MDL number MFCD05230713
  • Cas Number : 1157487-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69098

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69098
Availability details for
Quantity
  • Availability
  • Add to Cart