ENA308584626 Enamine

[(2-fluorophenyl)methyl]urea

  • MDL number MFCD01314571
  • Cas Number : 296277-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69101

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69101
Availability details for
Quantity
  • Availability
  • Add to Cart