ENA308584628 Enamine

4-chloro-N-cyclopropyl-2-nitroaniline

  • MDL number MFCD08448316
  • Cas Number : 304914-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70...72
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69102

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69102
Availability details for
Quantity
  • Availability
  • Add to Cart