ENA308584630 Enamine

N-ethyl-2,3-dihydro-1H-indole-5-carboxamide

  • MDL number MFCD11181011
  • Cas Number : 1094284-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69401

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69401
Availability details for
Quantity
  • Availability
  • Add to Cart