ENA308584637 Enamine

2-(dimethylamino)pyridin-3-ol

  • MDL number MFCD16998144
  • Cas Number : 233766-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69408
Availability details for
Quantity
  • Availability
  • Add to Cart