ENA308584639 Enamine

4-chloro-2-(1H-pyrrol-1-yl)aniline

  • MDL number MFCD12165072
  • Cas Number : 687638-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69411

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69411
Availability details for
Quantity
  • Availability
  • Add to Cart