ENA308584642 Enamine

1-(cyclopropylcarbamoyl)cyclobutane-1-carboxylic acid

  • MDL number MFCD16669513
  • Cas Number : 1250624-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69106

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69106
Availability details for
Quantity
  • Availability
  • Add to Cart