ENA308584644 Enamine

3-[(cyclobutylmethyl)amino]propanenitrile

  • MDL number MFCD11629602
  • Cas Number : 1095242-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69416

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69416
Availability details for
Quantity
  • Availability
  • Add to Cart