ENA308584649 Enamine

5-(dimethylamino)-4-(propan-2-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD09041342
  • Cas Number : 926193-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69428

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69428
Availability details for
Quantity
  • Availability
  • Add to Cart