ENA308584651 Enamine

N-(4-hydroxyphenyl)thiophene-3-carboxamide

  • MDL number MFCD11167683
  • Cas Number : 1042593-15-6
  • Molecular Weight : 219.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69430

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69430
Availability details for
Quantity
  • Availability
  • Add to Cart