ENA308584652 Enamine

3-[(2-oxo-2,3-dihydro-1H-indol-1-yl)methyl]furan-2-carboxylic acid

  • MDL number MFCD12652091
  • Cas Number : 1097140-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69109

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69109
Availability details for
Quantity
  • Availability
  • Add to Cart