ENA308584659 Enamine

(2-{4-[(1H-imidazol-1-yl)methyl]phenyl}phenyl)methanamine

  • MDL number MFCD09040730
  • Cas Number : 926237-60-7
  • Molecular Weight : 263.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69441

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69441
Availability details for
Quantity
  • Availability
  • Add to Cart