ENA308584663 Enamine

N1-[3-(1H-imidazol-1-yl)propyl]benzene-1,2-diamine

  • MDL number MFCD11196009
  • Cas Number : 262357-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69113

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69113
Availability details for
Quantity
  • Availability
  • Add to Cart