ENA308584664 Enamine

2-(benzylamino)-4-chlorobenzonitrile

  • MDL number MFCD12118969
  • Cas Number : 296259-54-6
  • Molecular Weight : 242.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69451

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69451
Availability details for
Quantity
  • Availability
  • Add to Cart