ENA308584666 Enamine

1,1-diphenylethan-1-amine

  • MDL number MFCD03002278
  • Cas Number : 20912-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69453

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69453
Availability details for
Quantity
  • Availability
  • Add to Cart