ENA308584672 Enamine

5-methylfuro[3,2-b]pyridine-2-carboxylic acid hydrochloride

  • MDL number MFCD11519398
  • Cas Number : 66497-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69462

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-69462
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart