ENA308584674 Enamine

N-[(1H-1,3-benzodiazol-2-yl)methyl]-N-methylpiperidin-4-amine trihydrochloride

  • MDL number MFCD17167247
  • Cas Number : 1252397-72-0
  • Molecular Weight : 353.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69114

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69114
Availability details for
Quantity
  • Availability
  • Add to Cart