ENA308584692 Enamine

(1,4-dioxan-2-yl)methanamine hydrochloride

  • MDL number MFCD06739050
  • Cas Number : 1185156-19-7
  • Molecular Weight : 153.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69487

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69487
Availability details for
Quantity
  • Availability
  • Add to Cart