ENA308584694 Enamine

1-[(dimethyl-1,3-oxazol-2-yl)methyl]piperazine

  • MDL number MFCD12773081
  • Cas Number : 1179018-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69119

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69119
Availability details for
Quantity
  • Availability
  • Add to Cart