ENA308584695 Enamine

3-[(pyridin-4-yl)amino]propan-1-ol

  • MDL number MFCD11124335
  • Cas Number : 68970-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69490

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69490
Availability details for
Quantity
  • Availability
  • Add to Cart