ENA308584697 Enamine

[2-(dimethylamino)ethyl][(3-methylphenyl)methyl]amine

  • MDL number MFCD09046178
  • Cas Number : 755008-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69120

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69120
Availability details for
Quantity
  • Availability
  • Add to Cart