ENA308584698 Enamine

ethyl 2-cyano-2-(1-methyl-2-oxo-2,3-dihydro-1H-indol-3-yl)acetate

  • MDL number MFCD17480422
  • Cas Number : 108272-71-5
  • Molecular Weight : 258.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69495

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69495
Availability details for
Quantity
  • Availability
  • Add to Cart