ENA308584699 Enamine

5-phenoxypyridin-2-amine

  • MDL number MFCD11135788
  • Cas Number : 64064-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67-69
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69121

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69121
Availability details for
Quantity
  • Availability
  • Add to Cart