ENA308584704 Enamine

1-(3-aminocyclohexyl)pyrrolidin-2-one

  • MDL number MFCD09806129
  • Cas Number : 1016531-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69502
Availability details for
Quantity
  • Availability
  • Add to Cart