ENA308584707 Enamine

1H-indole-4-carboxamide

  • MDL number MFCD09834878
  • Cas Number : 1670-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69507
Availability details for
Quantity
  • Availability
  • Add to Cart