ENA308584712 Enamine

4-[(1-methyl-1H-pyrazol-4-yl)methoxy]phenol

  • MDL number MFCD11181708
  • Cas Number : 1152531-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186...188
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69516

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69516
Availability details for
Quantity
  • Availability
  • Add to Cart