ENA308584716 Enamine

4-chloro-2-[(1-methylpiperidin-4-yl)amino]benzonitrile

  • MDL number MFCD12118943
  • Cas Number : 939816-96-3
  • Molecular Weight : 249.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69124
Availability details for
Quantity
  • Availability
  • Add to Cart