ENA308584720 Enamine

methyl 4-amino-3-fluorobenzoate

  • MDL number MFCD08689703
  • Cas Number : 185629-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111...113
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69530

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69530
Availability details for
Quantity
  • Availability
  • Add to Cart