ENA308584721 Enamine

4-chloro-2-(cyclohexylamino)benzonitrile

  • MDL number MFCD12118976
  • Cas Number : 939816-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69126

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69126
Availability details for
Quantity
  • Availability
  • Add to Cart