ENA308584729 Enamine

2-(5,6,7,8-tetrahydronaphthalene-2-amido)thiophene-3-carboxylic acid

  • MDL number MFCD13669604
  • Cas Number : 1099118-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-69549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69549
Availability details for
Quantity
  • Availability
  • Add to Cart