ENA308584739 Enamine

4-bromo-1-[2-(pyrrolidin-1-yl)ethyl]-1H-pyrazole

  • MDL number MFCD12824343
  • Cas Number : 951259-24-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69561

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69561
Availability details for
Quantity
  • Availability
  • Add to Cart