ENA308584740 Enamine

2-[2-(furan-2-yl)-1,3-thiazol-4-yl]acetamide

  • MDL number MFCD11644012
  • Cas Number : 1098349-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69562

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69562
Availability details for
Quantity
  • Availability
  • Add to Cart