ENA308584743 Enamine

N-{[2-(trifluoromethoxy)phenyl]methyl}cyclopentanamine

  • MDL number MFCD11139819
  • Cas Number : 1019579-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-69565

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69565
Availability details for
Quantity
  • Availability
  • Add to Cart