ENA308584748 Enamine

4-(cyclohexyloxy)phenol

  • MDL number MFCD11103426
  • Cas Number : 42873-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62-63
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69582

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69582
Availability details for
Quantity
  • Availability
  • Add to Cart