ENA308584755 Enamine

5-ethyl-4-(4-methoxyphenyl)-1,3-thiazol-2-amine

  • MDL number MFCD02232182
  • Cas Number : 493028-20-9
  • Molecular Weight : 234.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69596

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69596
Availability details for
Quantity
  • Availability
  • Add to Cart